Order Entry
Italy
ContactUsLinkComponent
Anti-Claudin 16 Rabbit Polyclonal Antibody
Anti-Claudin 16 Rabbit Polyclonal Antibody
Catalog # BOSSBS-13752R
Supplier:  Bioss
undefined
Anti-Claudin 16 Rabbit Polyclonal Antibody
Catalog # BOSSBS-13752R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Claudin 16
  • Antigen symbol:
    CLDN16
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Antigen synonyms:
    Cldn16|Paracellin-1|Paracellin 1|CLD16_HUMAN|PCLN1.|CLDN 16|Claudin 16|Claudin-16|PCLN-1,PCLN 1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Tight junctions mediate the regulation of the paracellular pathway between epithelial and endothelial cells. They form close connections to eliminate the extracellular space and regulate the flow of solutes between cells. The human gene PCLN-1 (paracellin-1) is related to the claudin family of integral membrane proteins, which localize to tight junctions. PCLN-1 contains four transmembrane domains and intracellular amino and carboxy termini, characteristic of the other claudin family members, and is detected only at the tight junctions of kidney tissue. PCLN-1 forms an intercellular pore and controls the resorption of magnesium and calcium in the thick ascending limb of Henle (TAL). Mutations in PCLN-1 cause renal magnesium wasting, which may contribute to a rare autosomal recessive disease, renal hypomagnesemia with hypercalciuria and nephrocalcinosis.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Claudin 16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat