Specifications
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 58
- Antigen symbol:GPR58
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Gene ID:9287
- Antigen synonyms:GPR 58|phBL5|G protein coupled receptor 58|TAAR2|Trace amine associated receptor 2.|GPCR 58
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Trace amines are endogenous molecules structurally related to classical biogenic amines that are linked to psychiatric conditions. A family of G-protein coupled receptors referred to as trace-amine-associated receptors (TAAR) are activated by trace amines and are present in very low levels in mammalian tissue. TAAR’s contain several structural features that are similar to the rhodopsin beta-adrenergic receptor superfamily, including the positions of the seven transmembrane regions that provide common ligand-binding pockets as well as the short N- and C-terminal domains. TAAR proteins are potential targets for drugs of abuse, such as amphetamine and MDMA, as well as neuropsychiatric disorders including schizophrenia, depression, and attention deficit disorder. TAAR-1 is a 340 amino acid protein that increases intracellular cAMP accumulation in response to beta-phenylethylamine and tyramine. TAAR-1 is associated with the detection of social cues, illustrating its significance as a therapeutic target.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: GPR58
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human