- Antibody type:Primary
- Antigen name:Nogo B Receptor
- Antigen symbol:NGBR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Antigen synonyms:nuclear undecaprenyl pyrophosphate synthase 1 homolog|NGBR_HUMAN.|NUS1|NGBR|C6orf68
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Nogo is an oligodendrocyte-specific member of the Reticulon family and is a component of CNS white matter that inhibits axon outgrowth, induces collapse of growth cones of chick dorsal root ganglion cells, and inhibits the spreading of 3T3 fibroblasts. Nogo is expressed by oligodendrocytes but not by Schwann cells and associates primarily with the endoplasmic reticulum. Nogo exists in three different splice forms, Nogo-A, -B and -C. NgBR (Nogo-B receptor), also known as nuclear undecaprenyl pyrophosphate synthase 1 homolog, is a 293 amino acid single-pass type I membrane protein that acts as a specific receptor for the amino-terminus of Nogo-B. Through this interaction, NgBR is involved in the regulation of vascular remodeling and angiogenesis. NgBR also enhances Niemann-Pick type C2 protein (NPC2) stabilization. Knockdown of NgBR mRNA leads to decreased NPC2 levels, which results in the hallmarks of NPC2 mutation: increased intracellular cholesterol accumulation and a loss of sterol sensing.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: NGBR/Nogo B receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat