Anti-IkB beta Rabbit Polyclonal Antibody
Catalog # BOSSBS-10246R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen symbol:IkB beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Antigen synonyms:IkappaBbeta|Thyroid receptor-interacting protein 9|IKBB_HUMAN|IkB-B|IKBB|IkB-beta|TR interacting protein 9|Nfkbib|NF kappa BIB|I kappa B beta|TRIP9.|IKB beta|I-kappa-B-beta|NF-kappa-B inhibitor beta|TR-interacting protein 9|IkBbeta|NF-kappa-BIB|Thyroid receptor interacting protein 9|TRIP-9
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
The protein encoded by this gene belongs to the NF-kappa-B inhibitor family, which inhibit NF-kappa-B by complexing with, and trapping it in the cytoplasm. Phosphorylation of serine residues on these proteins by kinases marks them for destruction via the ubiquitination pathway, thereby allowing activation of the NF-kappa-B, which translocates to the nucleus to function as a transcription factor. Alternatively spliced transcript variants have been found for this gene.[provided by RefSeq, Jul 2011].
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: IkB beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat