Order Entry
Italy
ContactUsLinkComponent
Anti-BMP4 Rabbit Polyclonal Antibody
Anti-BMP4 Rabbit Polyclonal Antibody
Catalog # BSBTPB9688
Supplier:  Boster Bio
undefined
Anti-BMP4 Rabbit Polyclonal Antibody
Catalog # BSBTPB9688
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    bone morphogenetic protein 4
  • Antigen symbol:
    BMP4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    652
  • Antigen synonyms:
    BMP2B1|MCOPS6|ZYME|OFC11|BMP2B
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Bone morphogenetic protein 4(BMP4) detection. Tested with WB, ELISA in Human;Mouse.

Type: Primary
Antigen: BMP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse