Order Entry
Italy
ContactUsLinkComponent
Anti-G6PD Rabbit Polyclonal Antibody
Anti-G6PD Rabbit Polyclonal Antibody
Catalog # ABNOPAB29349
Supplier:  Abnova
Anti-G6PD Rabbit Polyclonal Antibody
Catalog # ABNOPAB29349
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glucose-6-phosphate dehydrogenase
  • Antigen symbol:
    G6PD
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    2539
  • Antigen synonyms:
    G6PD1
  • Sequence:
    FQGDAFHQSDTHIFIIMGASGDLAKKKIYPTIWWLFRDGLLPENTFIVGYARSRLTVADIRKQSEPFFKATPEEKLKLEDFFARNSYVAGQYDDAASYQRLNSHMNALH
  • Immunogen:
    Recombinant protein corresponding to human G6PD.
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody raised against recombinant human G6PD.

Type: Primary
Antigen: G6PD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human