Order Entry
Italy
ContactUsLinkComponent
Anti-ITGAX Mouse Monoclonal Antibody [clone: 3.9] (FITC (Fluorescein Isothiocyanate))
Anti-ITGAX Mouse Monoclonal Antibody [clone: 3.9] (FITC (Fluorescein Isothiocyanate))
Catalog # ABNOMAB12219
Supplier:  Abnova
Anti-ITGAX Mouse Monoclonal Antibody [clone: 3.9] (FITC (Fluorescein Isothiocyanate))
Catalog # ABNOMAB12219
Supplier:  Abnova

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    integrin, alpha X (complement component 3 receptor 4 subunit)
  • Antigen symbol:
    ITGAX
  • Clonality:
    Monoclonal
  • Clone:
    3.9
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Gene ID:
    3687
  • Antigen synonyms:
    CD11C|SLEB6
  • Immunogen:
    Native purified from human rheumatoid synovial fluid cells and fibronectin purified human monocytes.
  • Size:
    100 Reactions
  • Pk:
    100 Assays

Specifications

About this item

Mouse monoclonal antibody raised against ITGAX.

Type: Secondary
Antigen: Native purified from human rheumatoid synovial fluid cells and fibronectin purified human monocytes.
Clonality: Monoclonal
Clone: 3.9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human