Anti-CD8A Rat Monoclonal Antibody [clone: 53-6.7] (APC (Allophycocyanin)/Cy7®)
ABNOMAB12215
: Abnova
- Antibody type:Secondary
- Antigen name:CD8 antigen, alpha chain
- Antigen symbol:CD8A
- Clonality:Monoclonal
- Clone:53-6.7
- Conjugation:APC (Allophycocyanin)/Cy7®
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Gene ID:12525
- Antigen synonyms:BB154331|Ly-2|Lyt-2|Ly-35|Ly-B
- Immunogen:Cd8a
- Size:100 µg
- Pk:100 µG
Rat monoclonal antibody raised against Cd8a.
Type: Secondary
Antigen: Cd8a
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: APC (Allophycocyanin)/Cy7
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse