Order Entry
Italy
ContactUsLinkComponent
Copper di-ammonium solution Cu(NH₄)₂-EDTA 0.1 mol/l, Titriplex®, Supelco®
Copper di-ammonium solution Cu(NH₄)₂-EDTA 0.1 mol/l, Titriplex®, Supelco®
Catalog # 1.05217.0500
Supplier:  Merck
CAS Number:  
Copper di-ammonium solution Cu(NH₄)₂-EDTA 0.1 mol/l, Titriplex®, Supelco®
Catalog # 1.05217.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Assay (Cu):
    0.098 - 0.102 mol/l
  • pH-value:
    9.7 - 10.3
  • Free EDTA:
    conforms
  • Free copper:
    conforms

Specification Test Results

About this item

 :
Density:1.02 g/cm3 (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient