Anti-5HT2B Receptor Antibody
Catalog # BSBTPB9600
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:5-hydroxytryptamine (serotonin) receptor 2B
- Antigen symbol:HTR2B
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3357
- Antigen synonyms:5-HT2B|5-HT(2B)
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for 5-hydroxytryptamine receptor 2B(HTR2B) detection. Tested with WB in Human;Rat.
Type: Primary
Antigen: 5HT2B Receptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat