Anti-DMD Antibody Pair
Catalog # ABNOH00001756-PW1
Supplier: Abnova
Specifications
- Antigen name:Dystrophin
- Antigen symbol:DMD
- Host:Multiple
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:1756
- Antigen synonyms:DXS230|MRX85|DXS164|BMD|DXS272|DXS268|DXS206|DXS142|CMD3B|DXS269|DXS270|DXS239
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-DMD (300 µg); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-DMD (50 µl)
Type:
Antigen: DMD
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human