Anti-PTBP1 Antibody Pair
Catalog # ABNOH00005725-PW1
Supplier: Abnova
Specifications
- Antigen name:Polypyrimidine tract binding protein 1
- Antigen symbol:PTBP1
- Host:Multiple
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:5725
- Antigen synonyms:PTB-1|PTB3|PTB-T|HNRNPI|HNRNP-I|PTB4|pPTB|PTB|HNRPI|PTB2
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-PTBP1 (300 µg); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-PTBP1 (50 µl)
Type:
Antigen: PTBP1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human