Order Entry
Italy
ContactUsLinkComponent
Anti-PRKAB1 Antibody Pair
Anti-PRKAB1 Antibody Pair
Catalog # ABNOH00005564-AP51
Supplier:  Abnova
Anti-PRKAB1 Antibody Pair
Catalog # ABNOH00005564-AP51
Supplier:  Abnova

Specifications

  • Antigen name:
    Protein Kinase, AMP-activated Non-catalytic Subunit Beta 1
  • Antigen symbol:
    PRKAB1
  • Host:
    Multiple
  • Reactivity:
    Human
  • Gene ID:
    5564
  • Antigen synonyms:
    kinase AMPK-beta1|AMPK-beta1|AMPK beta-1 chain|5'-AMP-activated protein kinase beta-1 subunit|PRKAB1|AAKB1
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

Specifications

About this item

This Set comes with matched Antibody Pair to detect and quantify protein level of human PRKAB1.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-PRKAB1 (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-PRKAB1 (50 µg);

Type:
Antigen: PRKAB1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human