VWR®, Aluminium foil in dispensers
Supplier: VWR Collection
Certificates
About this item
Pure aluminium 99,0 to 99,5%.
- Comes in a practical dispenser
- Available in various widths and thicknesses
- Roll length to suit various requirements
Ideal for packaging, covering and insulation.