Order Entry
Italy
ContactUsLinkComponent
Pycnometers, Hubbard
Pycnometers, Hubbard
Catalog # NEUB0303-50-25
Pycnometers, Hubbard
Catalog # NEUB0303-50-25
Specifications
  • NS:
    24/15
  • Description:
    Pycnometer, conical, unadjustable, 25 ml
  • Seq:
    02
  • Pk:
    1
Specifications
About this item
Borosilicate glass 3,3.
  • With glass stopper
  • For viscous liquids, pastes such as petroleum jelly, creams and bituminous masses
According to DIN 12806 and DIN ISO 3507.