Specifications
- Antibody type:Primary
- Antigen name:Peroxisome proliferator-activated receptor gamma
- Antigen symbol:PPAR gamma
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Isotype:IgY
- Reactivity:Human
- Western blot:Yes
- Epitope:Amino acids 115-129 of Human Peroxisome proliferator-activated receptor gamma
- Cross adsorption:No
- Format:Liquid
- Immunogen:Human Peroxisome proliferator-activated receptor gamma peptide (115-129 aa).
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Peroxisome proliferator-activated receptor gamma (PPARG) is a nuclear hormone receptor that binds peroxisome proliferators such as hypolipidemic drugs and fatty acids. Once activated by a ligand, the receptor binds to a promoter element in the gene for acyl-CoA oxidase and activates its transcription (Ref: SWISSPROT).
Antibody detects PPARG by western blot in human cell nuclear extracts. It is recommended to isolate nuclei from stimulated cultures for best results.
Application Information:
WB. Suggested dilution at 1:2,000 to 1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: Peroxisome proliferator-activated receptor gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino acids 115-129 of Human Peroxisome proliferator-activated receptor gamma
Host: Chicken
Isotype: IgY
Reactivity: Human