Order Entry
Italy
ContactUsLinkComponent
Anti-Beta Endorphin Rabbit Polyclonal Antibody
Anti-Beta Endorphin Rabbit Polyclonal Antibody
Catalog # BSENR-1492-50
Supplier:  Biosensis
undefined
Anti-Beta Endorphin Rabbit Polyclonal Antibody
Catalog # BSENR-1492-50
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Beta Endorphin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Epitope:
    Multiple epitopes because of the nature of the immunogen
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Immunogen:
    Synthetic human beta-endorphin peptide (1-31 aa) conjugated to BSA.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Human beta-endorphin is a 31 amino acid peptide cleaved from the precursor pro-opiomelanocortin (POMC). It is an endogenous opioid peptide neurotransmitter that interacts with opioid receptors.

Confirmed by RIA against the synthetic peptide.
Application Information:
A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. For radioimmunoassay (RIA), a working dilution of 1:10,000 is recommended. Sensitivity - 120 pg/sample; IC50-1 ng/ sample. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Beta Endorphin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple epitopes because of the nature of the immunogen
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat