Order Entry
Italy
ContactUsLinkComponent
Anti-alpha synuclein Sheep Polyclonal Antibody
Anti-alpha synuclein Sheep Polyclonal Antibody
Catalog # BSENS-075-50
Supplier:  Biosensis
undefined
Anti-alpha synuclein Sheep Polyclonal Antibody
Catalog # BSENS-075-50
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Synuclein alpha
  • Antigen symbol:
    SNCA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Sheep
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    aa: 116-131
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Antigen synonyms:
    PARK1|PARK4|NACP|PD1
  • Immunogen:
    A synthetic peptide (CMPVDPDNEAYEMPSEE) of human alpha synuclein protein (aa: 116-131) conjugated to diphtheria toxoid has been used as the immunogen.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Alpha synuclein is an abundant 140 amino acid neuronal protein, expressed primarily at presynaptic terminals in the central nervous system. Alpha synuclein has been associated with several neurodegenerative diseases. A point mutation in the gene coding for the alpha-synuclein protein was the first discovery linking this protein to a rare familial form of Parkinson's disease (PD). Subsequently, other mutations in the alpha-synuclein gene have been identified in familial PD. The aggregated proteinaceous inclusions called Lewy bodies found in PD and cortical Lewy body dementia (LBD) were discovered to be predominantly alpha-synuclein. Aberrant aggregation of alpha-synuclein has been detected in an increasing number of neurodegenerative diseases, collectively known as synucleopathies. Alpha-synuclein exists physiologically in both soluble and membrane-bound states, in unstructured and alpha-helical conformations, respectively. The physiological function of alpha-synuclein appears to require its translocation between these subcellular compartments and interconversion between the 2 conformations. Abnormal processing of alpha-synuclein is predicted to lead to pathological changes in its binding properties and function.

This antiserum specifically detects alpha synuclein.
Application Information:
IHC, WB, ELISA. Recommended to be used at a concentration of 1 µg/ml for immunohistochemistry (Paraffin sections); 0.5 µg/ml for WB and immunoblot. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: alpha synuclein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: aa: 116-131
Host: Sheep
Isotype: IgG
Reactivity: Rat, Human, Mouse