Order Entry
Italy
ContactUsLinkComponent
Anti-GRIN1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: S308-48]
Anti-GRIN1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: S308-48]
Catalog # ABNOMAB11722
Supplier:  Abnova
Anti-GRIN1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: S308-48]
Catalog # ABNOMAB11722
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glutamate [NMDA] receptor subunit zeta-1
  • Antigen symbol:
    GRIN1
  • Clonality:
    Monoclonal
  • Clone:
    S308-48
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    24408
  • Antigen synonyms:
    NMDAR1|NMD-R1|MRD8|GluN1|NR1|NMDA1
  • Amino acid number:
    42 to 361
  • Storage buffer:
    PBS pH 7,4 (50% glycerol and 0,09% sodium azide).
  • Storage temperature:
    Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant fusion protein corresponding to amino acids 42-361 at extracellular N-terminus of rat Grin1.
  • Purification:
    Protein G purification
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against partial synthetic protein of Grin1.

Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: GRIN1
Clonality: Monoclonal
Clone: S308-48
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: