Order Entry
Italy
ContactUsLinkComponent
Anti-SCNN1B Rabbit Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))
Anti-SCNN1B Rabbit Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))
Catalog # ABNOPAB28799
Supplier:  Abnova
Anti-SCNN1B Rabbit Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))
Catalog # ABNOPAB28799
Supplier:  Abnova

Specifications

  • Antigen name:
    Sodium Channel Nonvoltage-gated 1 Beta
  • Antigen symbol:
    SCNN1B
  • Clonality:
    Polyclonal
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    24767
  • Antigen synonyms:
    ENaCb|BESC1|ENaCbeta|SCNEB
  • Storage buffer:
    PBS, 50% glycerol and 0,09% sodium azide
  • Storage temperature:
    Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to Scnn1b C-terminus (amino acids 617-638).
  • Purification:
    Affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against recombinant Rat Scnn1b.

Recommended Dilutions: Immunofluorescence Immunohistochemistry Immunoprecipitation Western Blot: 1:1000

Type:
Antigen: SCNN1B
Clonality: Polyclonal
Clone:
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rabbit
Isotype:
Reactivity: