Order Entry
Italy
ContactUsLinkComponent
Anti-MCM2 Mouse Monoclonal Antibody [clone: 2B3]
Anti-MCM2 Mouse Monoclonal Antibody [clone: 2B3]
Catalog # ABNOMAB10805
Supplier:  Abnova
Anti-MCM2 Mouse Monoclonal Antibody [clone: 2B3]
Catalog # ABNOMAB10805
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DNA replication licensing factor MCM2
  • Antigen symbol:
    MCM2
  • Clonality:
    Monoclonal
  • Clone:
    2B3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Monkey,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Ascites
  • Gene ID:
    4171
  • Antigen synonyms:
    MITOTIN|minichromosome maintenance protein 2 homolog|cyclin-like 1|D3S3194|cell devision cycle-like 1|CCNL1|MCM2 minichromosome maintenance deficient 2|mitotin|minichromosome maintenance deficient 2 (mitotin)|DNA replication licensing factor MCM2|nuclear protein BM28|BM28|cdc19|CDCL1
  • Storage buffer:
    0,03% sodium azide
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to human MCM2.
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant MCM2.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: MCM2
Clonality: Monoclonal
Clone: 2B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat