Anti-KDM3A Mouse Monoclonal Antibody [clone: 1E12]
Catalog # ABNOMAB10706
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Lysine (K)-specific Demethylase 3A
- Antigen symbol:KDM3A
- Clonality:Monoclonal
- Clone:1E12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:55818
- Antigen synonyms:JHDM2A|TSGA|JHMD2A|JMJD1A|JMJD1
- Storage buffer:0,03% sodium azide
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human JMJD1A.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant JMJD1A.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000
Type: Primary
Antigen: KDM3A
Clonality: Monoclonal
Clone: 1E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human