Anti-MKI67 Mouse Monoclonal Antibody [clone: 9C12B2]
Catalog # ABNOMAB10600
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Marker of proliferation Ki-67
- Antigen symbol:MKI67
- Clonality:Monoclonal
- Clone:9C12B2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:4288
- Antigen synonyms:MIB-1|MIB-|KIA|PPP1R105
- Amino acid number:3118 to 3256
- Storage buffer:0,03% sodium azide
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 3118-3256 of human MKI67.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant MKI67.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000
Type: Primary
Antigen: MKI67
Clonality: Monoclonal
Clone: 9C12B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human