Anti-CA9 Mouse Monoclonal Antibody [clone: 2D3]
Catalog # ABNOMAB10517
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Carbonic Anhydrase 9
- Antigen symbol:CA9
- Clonality:Monoclonal
- Clone:2D3
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:768
- Antigen synonyms:CAIX|carbonic dehydratase|membrane antigen MN|carbonic anhydrase 9|RCC-associated protein G250|MN|carbonate dehydratase IX|CA-IX|RCC-associated antigen G250|P54/58N|pMW1|renal cell carcinoma-associated antigen G250
- Storage buffer:0,03% sodium azide
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human CA9.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant CA9.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Flow Cytometry: 1:200-1:400
Type: Primary
Antigen: CA9
Clonality: Monoclonal
Clone: 2D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human