Anti-CK7 Mouse Monoclonal Antibody
Catalog # ABNOMAB11153
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin 7
- Antigen symbol:CK7
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:3855
- Antigen synonyms:CK7|SCL|K2C7|K7
- Storage buffer:PBS, pH 7,4 (1% BSA, 50% glycerol, 0,02% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to N-terminus of human KRT7.
- Purification:Protein G purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant protein of KRT7.
Recommended Dilutions: ELISA: 1:5000-1:20000; Western Blot: 1:100-1:2000
Type: Primary
Antigen: CK7
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human