Anti-GCC1 Mouse Polyclonal Antibody
Catalog # ABNOH00079571-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:GRIP and coiled-coil domain containing 1
- Antigen symbol:GCC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:79571
- Antigen synonyms:GCC1_HUMAN.|Golgi coiled coil protein 1|GCC protein|Golgi coiled coil 1|Peripheral membrane Golgi protein|GCC88|GRIP and coiled coil domain containing 1|GCC1P
- Amino acid number:1 to 775
- Storage buffer:1x PBS, pH 7,4
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:GCC1 (NP_078799.2, 1 a.a. ~ 775 a.a) full-length human protein.
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Mouse polyclonal antibody raised against a full-length human GCC1 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: GCC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human