Order Entry
Italy
ContactUsLinkComponent
 
Anti-TCIRG1 Mouse Monoclonal Antibody [clone: 7H20]
  ABNOH00010312-M02A
 :  Abnova
Anti-TCIRG1 Mouse Monoclonal Antibody [clone: 7H20]
  ABNOH00010312-M02A
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    T-cell, immune Regulator 1, ATPase, H+ transporting, lysosomal V0 Subunit A3
  • Antigen symbol:
    TCIRG1
  • Clonality:
    Monoclonal
  • Clone:
    7H20
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Ascites fluid
  • Gene ID:
    10312
  • Antigen synonyms:
    V-type proton ATPase 116 kDa subunit a isoform 3|TIRC7|OPTB1|ATP6N1C|OC116|Osteoclastic proton pump 116 kDa subunit|Stv1|OC-116kDa|a3|V-ATPase 116 kDa isoform a3|Atp6i|Vph1|ATP6V0A3|TCIRG1
  • Amino acid number:
    121 to 220
  • Sequence:
    QLHQLQLHAAVLRQGHEPQLAAAHTDGASERTPLLQAPGGPHQDLRVNFVAGAVEPHKAPALERLLWRACRGFLIASFRELEQPLEHPVTGEPATWMTFL
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    TCIRG1 (AAH18133, 121 a.a. ~ 220 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
  • Size:
    200 μl
  • Pk:
    200 µl

 

 

Mouse monoclonal antibody raised against a partial recombinant TCIRG1.

Type: Primary
Antigen: TCIRG1
Clonality: Monoclonal
Clone: 7H20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human