Anti-CD53 Mouse Monoclonal Antibody [clone: MEM-53]
ABNOMAB0890
: Abnova
- Antibody type:Primary
- Antigen name:Leukocyte surface antigen CD53
- Antigen symbol:CD53
- Clonality:Monoclonal
- Clone:MEM-53
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:963
- Antigen synonyms:MOX44|TSPAN25|CD53 molecule
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified CD53 from leukocytes of a patient suffering from a LGL-type leukemia.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against native CD53.
Type: Primary
Antigen: CD53
Clonality: Monoclonal
Clone: MEM-53
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human