Order Entry
Italy
ContactUsLinkComponent
Anti-HLA-DPB1 Mouse Monoclonal Antibody [clone: HL-38]
Anti-HLA-DPB1 Mouse Monoclonal Antibody [clone: HL-38]
Catalog # ABNOMAB0900
Supplier:  Abnova
Anti-HLA-DPB1 Mouse Monoclonal Antibody [clone: HL-38]
Catalog # ABNOMAB0900
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HLA class II histocompatibility antigen, DP(W2) beta chain
  • Antigen symbol:
    HLA-DPB1
  • Clonality:
    Monoclonal
  • Clone:
    HL-38
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Pig,
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    3115|3123|3125|3126|3127
  • Antigen synonyms:
    DPB1|HLA-DP|HLA-DPB|HLA-DP1B
  • Storage buffer:
    PBS, pH 7,4 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. Do not freeze. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified HLA-DR/HLA-DP from Raji human Burkitt's lymphoma cell line.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against native HLA-DR/HLA-DP.

Type: Primary
Antigen: HLA-DPB1
Clonality: Monoclonal
Clone: HL-38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Pig