Order Entry
Italy
ContactUsLinkComponent
 
Anti-GAL4 Mouse Monoclonal Antibody [clone: 8C-1]
  ABNOMAB1884
 :  Abnova
Anti-GAL4 Mouse Monoclonal Antibody [clone: 8C-1]
  ABNOMAB1884
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen symbol:
    GAL4
  • Clonality:
    Monoclonal
  • Clone:
    8C-1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Yeast
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    855828
  • Storage buffer:
    PBS (0,08% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to full length yeast GAL4.
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against full length recombinant GAL4.

Recommended Dilutions: Western Blot: 1-10 ?g/ml

Type: Primary
Antigen: GAL4
Clonality: Monoclonal
Clone: 8C-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Yeast