Anti-CER1 Mouse Monoclonal Antibody [clone: 9D6]
ABNOMAB10470
: Abnova
- Antibody type:Primary
- Antigen name:Cerberus 1, DAN family BMP antagonist
- Antigen symbol:CER1
- Clonality:Monoclonal
- Clone:9D6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:9350
- Antigen synonyms:CER1|MGC119895|DAND4|MGC119894|MGC96951
- Storage buffer:ascitic (0,03% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human CER1.
- Size:100 μl
- Pk:100 µl
Mouse monoclonal antibody raised against recombinant CER1.
Recommended Dilutions: Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; ELISA: 1:10000
Type: Primary
Antigen: CER1
Clonality: Monoclonal
Clone: 9D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human