Anti-CD3 Monoclonal Antibody [clone: HCD3-D2]
Catalog # ABNOMAB1362
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD3 Molecule
- Antigen symbol:CD3
- Clonality:Monoclonal
- Clone:HCD3-D2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Isotype:IgG2
- Reactivity:Human
- Cross adsorption:No
- Format:Lyophilized
- Storage buffer:PBS
- Storage temperature:Store at –20 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified CD3 from peripheral blood T cells.
- Size:200 μg
- Pk:200 µg
Specifications
About this item
Mouse monoclonal antibody raised against native CD3.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: HCD3-D2
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2
Reactivity: Human