Anti-PRKCG Mouse Monoclonal Antibody [clone: M110]
Catalog # ABNOMAB1367
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase C Gamma
- Antigen symbol:PRKCG
- Clonality:Monoclonal
- Clone:M110
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:5582
- Antigen synonyms:PKCG|SCA14|PKCC|MGC57564|PKC-gamma
- Amino acid number:499 to 697
- Storage buffer:PBS (50% glycerol, 1 mg/mL BSA, 0,05% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 499-697 of human PRKCG.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant PRKCG.
Recommended Dilutions: ELISA: 1:2000; Immunocytochemistry: 1:100; Immunoprecipitation: 1-5 ?l; Western Blot: 1:1000
Type: Primary
Antigen: PRKCG
Clonality: Monoclonal
Clone: M110
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat