Order Entry
Italy
ContactUsLinkComponent
Anti-CCNA2 Mouse Monoclonal Antibody [clone: CY-28]
Anti-CCNA2 Mouse Monoclonal Antibody [clone: CY-28]
Catalog # ABNOMAB1448
Supplier:  Abnova
Anti-CCNA2 Mouse Monoclonal Antibody [clone: CY-28]
Catalog # ABNOMAB1448
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclin A2
  • Antigen symbol:
    CCNA2
  • Clonality:
    Monoclonal
  • Clone:
    CY-28
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Bovine,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Gene ID:
    281667
  • Antigen synonyms:
    CCN1|CCNA|CCNA2|Cyclin A|Cyclin-A|CyclinA|Cyclin-A2
  • Storage buffer:
    1.2% sodium acetate (2 mg BSA, 0,01 mg sodium azide)
  • Storage temperature:
    Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to full length bovine CCNA2.
  • Purification:
    Affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against full length recombinant CCNA2.

Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: CCNA2
Clonality: Monoclonal
Clone: CY-28
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Human, Mouse