Anti-HSPD1 Mouse Monoclonal Antibody [clone: SJ-60]
Catalog # ABNOMAB1460
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Heat shock 60kDa protein 1
- Antigen symbol:HSPD1
- Clonality:Monoclonal
- Clone:SJ-60
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Chicken,Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Lyophilized
- Gene ID:3329
- Antigen synonyms:GROEL|HSP65|HLD4|HSP-60|HuCHA60|Chaperonin 60|HSP60|Mitochondrial Matrix Protein P1|CPN60|Heat Shock Protein 65|SPG13
- Storage buffer:1.2% sodium acetate (2 mg BSA, 0,01 mg sodium azide)
- Storage temperature:Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human HSPD1.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant HSPD1.
Recommended Dilutions: Western Blot: 2-4 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 4-8 ?g/ml
Type: Primary
Antigen: HSPD1
Clonality: Monoclonal
Clone: SJ-60
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Rat