Anti-S100 A11 Mouse Monoclonal Antibody [clone: YK-1A4]
ABNOMAB10144
: Abnova
- Antibody type:Primary
- Antigen name:S100 Calcium Binding Protein A11
- Antigen symbol:S100A11
- Clonality:Monoclonal
- Clone:YK-1A4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:6282
- Antigen synonyms:MLN70|HEL-S-43|S100C|Calgizzarin|Protein S100C|Protein S100-A11|S100A11|S100 calcium-binding protein A11
- Storage buffer:20 mM PBS, pH 7,5, 300 mM NaCl (50% glycerol)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human S100A11.
- Purification:Protein A purification
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against full length recombinant S100A11.
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: S100 A11
Clonality: Monoclonal
Clone: YK-1A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human