Order Entry
Italy
ContactUsLinkComponent
IgG1 Isotype Control [clone: MOPC-21] (PE (Phycoerythrin))
IgG1 Isotype Control [clone: MOPC-21] (PE (Phycoerythrin))
Catalog # ABNOMAB10156
Supplier:  Abnova
IgG1 Isotype Control [clone: MOPC-21] (PE (Phycoerythrin))
Catalog # ABNOMAB10156
Supplier:  Abnova

Specifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    MOPC-21
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS (0,2% BSA, 15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. Do not freeze.
  • Purification:
    Size-exclusion chromatography purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody was chosen by screening on variety of resting, activated, live and fixed rat and human tissues.

Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: PE (Phycoerythrin)
Epitope:
Host:
Isotype: IgG1
Reactivity: