Anti-SYP Mouse Monoclonal Antibody [clone: SYN-58]
Catalog # ABNOMAB1527
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Synaptophysin
- Antigen symbol:SYP
- Clonality:Monoclonal
- Clone:SYN-58
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Lyophilized
- Gene ID:24804
- Antigen synonyms:MRXSYP|major synaptic vesicle protein P38|MRX96
- Storage buffer:1.2% sodium acetate (2 mg BSA, 0,01 mg sodium azide)
- Storage temperature:Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified rat Syp.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against native Syp.
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; Immunohistochemistry (Frozen sections): 1-2 ?g/ml
Type: Primary
Antigen: SYP
Clonality: Monoclonal
Clone: SYN-58
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat