Anti-KLK5 Rabbit Polyclonal Antibody
Catalog # ABNOPAB0138
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Kallikrein-related peptidase 5
- Antigen symbol:KLK5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Lyophilized
- Form:Serum
- Storage temperature:Store at –20 °C on dry atmosphere. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A mixture of synthetic peptides (conjugated to KLH) corresponding to KLK5.
- Size:200 μl
- Pk:200 µl
Specifications
About this item
Rabbit polyclonal antibody raised against a mixture of synthetic peptide conjugated to KLH.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: KLK5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human