Anti-NCOA3 Rabbit Polyclonal Antibody
Catalog # ABNOPAB10282
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Nuclear receptor coactivator 3
- Antigen symbol:NCOA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Macaque,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:8202
- Antigen synonyms:SRC3|RAC3|TNRC16|TNRC14|CAGH16|MGC141848|KAT13B|SRC-3|AIB1|TRAM-1|AIB-1|pCIP|bHLHe42|CTG26|ACTR|RP5-1049G16.3
- Storage buffer:20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal region of human NCOA3.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of NCOA3.
Recommended Dilutions: ELISA: 1:175000; Western Blot: 1:500-1:1000
Type: Primary
Antigen: NCOA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Human, Macaque, Mouse, Rat