Anti-RSL1D1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB10306
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ribosomal L1 Domain Containing 1
- Antigen symbol:RSL1D1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:26156
- Antigen synonyms:RSL1D1|MGC142259|cellular senescence inhibited gene protein|ribosomal L1 domain containing 1|CSIG|L12|DKFZP564M182|PBK1|MGC138433
- Storage buffer:20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal region of human RSL1D1.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of RSL1D1.
Recommended Dilutions: ELISA: 1:2500-1:10000; Western Blot: 1:500-1:2000; Immunoprecipitation: 1:100
Type: Primary
Antigen: RSL1D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human