Anti-CHAF1B Mouse Monoclonal Antibody [clone: SS 24 1-68]
Catalog # ABNOMAB2354
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Chromatin Assembly Factor 1, Subunit B (p60)
- Antigen symbol:CHAF1B
- Clonality:Monoclonal
- Clone:SS 24 1-68
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Primates,Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:8208
- Antigen synonyms:CAF-1|MPHOSPH7|CAF1P60|CAF-IP60|MPP7|CAF1A|CAF1
- Storage buffer:buffer containing 0,1% sodium azide
- Storage temperature:Store at –20 °C or –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human CHAF1B.
- Size:50 μl
- Pk:50 µl
Specifications
About this item
Mouse monoclonal antibody raised against recombinant CHAF1B.
Recommended Dilutions: Immunoprecipitation: 0.1-1 ?g/ml; Western Blot: 1:1000
Type: Primary
Antigen: CHAF1B
Clonality: Monoclonal
Clone: SS 24 1-68
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Primates