Order Entry
Italy
ContactUsLinkComponent
Anti-H2-D2 Mouse Monoclonal Antibody [clone: 27-11-13S]
Anti-H2-D2 Mouse Monoclonal Antibody [clone: 27-11-13S]
Catalog # ABNOMAB3261
Supplier:  Abnova
Anti-H2-D2 Mouse Monoclonal Antibody [clone: 27-11-13S]
Catalog # ABNOMAB3261
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    H2-D2
  • Clonality:
    Monoclonal
  • Clone:
    27-11-13S
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    100 mM BBS, pH 8,2
  • Storage temperature:
    Store at 4 °C.
  • Immunogen:
    Native from C3H alphaBDF1 splenocytes.
  • Size:
    500 μg
  • Pk:
    500 µg

Specifications

About this item

Mouse monoclonal antibody raised against H2-D2.

Type: Primary
Antigen: H2-D2
Clonality: Monoclonal
Clone: 27-11-13S
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse