Anti-MKI67 Rabbit Polyclonal Antibody
ABNOPAB12383
: Abnova
- Antibody type:Primary
- Antigen name:Marker of proliferation Ki-67
- Antigen symbol:MKI67
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:4288
- Antigen synonyms:MIB-1|MIB-|KIA|PPP1R105
- Amino acid number:1200 to 1300
- Storage buffer:Tris-citrate/phosphate buffer, pH 7.0-8.0 (0,1% sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:A synthetic peptide corresponding to amino acids 1200-1300 of human MKI67.
- Pk:100 µl
Rabbit polyclonal antibody raised against synthetic peptide of MKI67.
Recommended Dilutions: Immunocytochemistry: 1:50-1:200; Immunofluorescence: 1:50-1:200; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:3200
Type: Primary
Antigen: MKI67
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human