Anti-RPTOR Rabbit Polyclonal Antibody
Catalog # ABNOPAB13419
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Regulatory associated protein of MTOR
- Antigen symbol:RPTOR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:57521
- Antigen synonyms:mKIAA1303|Raptor|Regulatory-associated protein of mTOR|4932417H02Rik|Rap
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal region 15 amino acids of human RPTOR.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of RPTOR.
Recommended Dilutions: Western Blot: 2 ?g/ml
Type: Primary
Antigen: RPTOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse