Order Entry
Italy
ContactUsLinkComponent
 
Anti-JAK2 Rabbit Polyclonal Antibody
  ABNOPAB12769
 :  Abnova
Anti-JAK2 Rabbit Polyclonal Antibody
  ABNOPAB12769
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Janus kinase 2
  • Antigen symbol:
    JAK2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    3717|16452|24514
  • Antigen synonyms:
    JTK 10|JAK-2|JAK2|Janus Kinase 2|JTK10|JAK 2|Janus Activating Kinase 2
  • Storage buffer:
    PBS, pH 7,2 (10% Proclin300)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to C-terminus of human, mouse and rat JAK2.
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Rabbit polyclonal antibody raised against synthetic peptide of JAK2.

Recommended Dilutions: Western Blot: 0.1-1 ?g/ml; ELISA: 0.01-0.1 ?g/ml; Immunoprecipitation: 2-5 ?g/ml; Immunohistochemistry: 1-2.5 ?g/ml

Type: Primary
Antigen: JAK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat