Order Entry
Italy
ContactUsLinkComponent
 
Anti-CD300E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: UP-H2]
Catalog # ABNOMAB4998
Supplier:  Abnova
Anti-CD300E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: UP-H2]
Catalog # ABNOMAB4998
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD300e molecule
  • Antigen symbol:
    CD300E
  • Clonality:
    Monoclonal
  • Clone:
    UP-H2
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    342510
  • Antigen synonyms:
    CLM2|IREM-2|PIgR-2|CMRF35-A5|IREM2|CD300LE|PIgR2|CLM-2
  • Storage buffer:
    buffer containing 1% BSA, pH 7,2 (0,09% sodium azide)
  • Storage temperature:
    Store in the dark at 4 °C. Avoid prolonged exposure to light.
  • Immunogen:
    Human monocytes CD300E.
  • Size:
    100 Reactions
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against CD300E.

Recommended Dilutions: Flow Cytometry: 20 ?l/10^6 cells

Type: Primary
Antigen: CD300E
Clonality: Monoclonal
Clone: UP-H2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human