Order Entry
Italy
ContactUsLinkComponent
Anti-CD58 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MEM-63]
Anti-CD58 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MEM-63]
Catalog # ABNOMAB5006
Supplier:  Abnova
Anti-CD58 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MEM-63]
Catalog # ABNOMAB5006
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD58 Molecule
  • Antigen symbol:
    CD58
  • Clonality:
    Monoclonal
  • Clone:
    MEM-63
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Pig,
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    965
  • Antigen synonyms:
    LFA-3|LFA3|ag3
  • Storage buffer:
    PBS (0,2% BSA, 15 mM sodium azide)
  • Storage temperature:
    Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified CD58 from NALM-6 human pre-B cell line.
  • Size:
    100 Reactions
  • Pk:
    100 Tests

Specifications

About this item

Mouse monoclonal antibody raised against native CD58.

Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: CD58
Clonality: Monoclonal
Clone: MEM-63
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig