Anti-CD59 Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-43]
Codice catalogo ABNOMAB5008
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD59 Molecule, Complement Regulatory Protein
- Antigen symbol:CD59
- Clonality:Monoclonal
- Clone:MEM-43
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:966
- Antigen synonyms:MIRL|HRF-20|MEM43|1F5|MACIF|MIN3|EJ16|MSK21|EL32|HRF20|p18-20|MIC11|MIN1|16.3A5|MIN2|MAC-IP|G344|EJ30
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified CD59 from thymocytes and T lymphocytes.
- Size:100 Reactions
- Pk:100 Tests
Specifications
About this item
Mouse monoclonal antibody raised against native CD59.
Recommended Dilutions: Flow Cytometry: 10 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)
Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: MEM-43
Conjugation: APC (Allophycocyanin)
Epitope:
Host:
Isotype: IgG2a
Reactivity: Human