Anti-TFRC Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-75]
Catalog # ABNOMAB5013
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Transferrin Receptor
- Antigen symbol:TFRC
- Clonality:Monoclonal
- Clone:MEM-75
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:7037
- Antigen synonyms:TRFR|T9|TFR1|CD71|p90|TFR|TR
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified TFRC from NALM-6 human pre-B cell line.
- Size:100 Reactions
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native TFRC.
Recommended Dilutions: Flow Cytometry: 10 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)
Type: Primary
Antigen: TFRC
Clonality: Monoclonal
Clone: MEM-75
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human